missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Xylylene dibromide, 97%, Thermo Scientific Chemicals

$48.93 - $365.23

Chemical Identifiers

CAS 91-13-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000175
InChI Key KGKAYWMGPDWLQZ-UHFFFAOYSA-N
Synonym 1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776
PubChem CID 66665
IUPAC Name 1,2-bis(bromomethyl)benzene
SMILES C1=CC=C(C(=C1)CBr)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1724106
View Documents
Thermo Scientific Chemicals
A1724106
5 g N/A
 
AAA1724114
View Documents
Thermo Scientific Chemicals
A1724114
25 g N/A
 
AAA1724122
View Documents
Thermo Scientific Chemicals
A1724122
100 g N/A
 
Description

Description

1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(bromomethyl)benzene is used in the synthesis of isothioureas involved in the inhibition of human nitric oxide synthases. Also used in the synthesis of tridentate carbene ligands.

Solubility
Solubility in toluene is almost transparent and ethanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

91-13-4
263.96
KGKAYWMGPDWLQZ-UHFFFAOYSA-N
66665
C1=CC=C(C(=C1)CBr)CBr
C8H8Br2
MFCD00000175
1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776
1,2-bis(bromomethyl)benzene

Specifications

91-13-4
1.96
MFCD00000175
UN3261
1,2-bis bromomethyl benzene, o-xylylene dibromide, alpha,alpha'-dibromo-o-xylene, benzene, 1,2-bis bromomethyl, o-bis bromomethyl benzene, o-xylylene bromide, 1,2-dibenzyl bromide, alpha,alpha'-dibromo-o-xylol, a,a'-dibromo-o-xylene, ccris 1776
C1=CC=C(C(=C1)CBr)CBr
263.96
263.96
o-Xylylene dibromide
92°C to 96°C
C8H8Br2
5 g
637159
KGKAYWMGPDWLQZ-UHFFFAOYSA-N
1,2-bis(bromomethyl)benzene
66665
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 202-042-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only