Oberon™ Splash Hood
Full head protection from intermittent liquid splash
Supplier: Oberon™ CS8AFR
Includes: Vinyl fabric hood, Face-Fit chemical-resistance polycarbonate window with ratchet headgear
Description
- Shell is designed with a full shape to protect the head, neck and shoulders
- Chemical-resistant coating on outer surface of window matches chemical/splash resistance of vinyl coated fabric
- Face-Fit™ chemical-resistant polycarbonate Window coated with Silicote™-6 resists damage caused by many mild acids, bases and solvents (except hydrofluoric acid)
- Inside of the window is permanent antifog coated for clear visibility and no fogging
- Includes ratchet headgear for easy adjustment
- Made in the USA
Certifications
Window meets American National Standards Institute (ANSI) regulation Z87.1-2010
Specifications
Anti-fog | |
Splash Hood | |
Vinyl |
ANSI Z87.1-2010 | |
Vinyl coated fabric hood; Polycarbonate window | |
Polycarbonate |
Will not wipe off hydrofluoric acid