missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octadecane 99.5+%, TCI America™
Supplier: TCI America S02905ML
Specifications
Octadecane [Standard Material for GC] | |
White | |
C18H38 | |
5 mL | |
RZJRJXONCZWCBN-UHFFFAOYSA-N | |
octadecane | |
11635 | |
254.50 | |
Crystalline Lumps |
593-45-3 | |
317°C | |
MFCD00009007 | |
n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group | |
CCCCCCCCCCCCCCCCCC | |
254.502 | |
CHEBI:32926 | |
≥99.5% (GC) |
Chemical Identifiers
593-45-3 | |
254.502 | |
RZJRJXONCZWCBN-UHFFFAOYSA-N | |
11635 | |
octadecane |
C18H38 | |
MFCD00009007 | |
n-octadecane, octadecan, unii-n102p6haiu, n102p6haiu, octadecane, octadecane, analytical standard, octadecane, n, oktadekan, ccris 681, stearyl group | |
CHEBI:32926 | |
CCCCCCCCCCCCCCCCCC |
Safety and Handling
EINECSNumber : (2)-0010
TSCA : Yes