missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octadecylamine, 90%, tech., Thermo Scientific Chemicals

$92.09 - $153.72

Chemical Identifiers

CAS 124-30-1
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.52
MDL Number MFCD00008159
InChI Key REYJJPSVUYRZGE-UHFFFAOYSA-N
Synonym octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
PubChem CID 15793
ChEBI CHEBI:63866
IUPAC Name octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC129325000
SDS View Documents
Thermo Scientific Chemicals
129325000
500 g Glass bottle
Each for $92.09
 
AC129320010
SDS View Documents
Thermo Scientific Chemicals
129320010
1 kg Glass bottle
Each for $153.72
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-30-1
269.52
REYJJPSVUYRZGE-UHFFFAOYSA-N
15793
octadecan-1-amine
C18H39N
MFCD00008159
octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
CHEBI:63866
CCCCCCCCCCCCCCCCCCN

Specifications

124-30-1
100.0
White
148°C
88% min. (GC)
C18H39N
MFCD00008159
04, IV, 825
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons
CCCCCCCCCCCCCCCCCCN
269.52
4.17 mPa.s (20°C)
269.51
Technical
Prills
88.0
50.0°C to 60.0°C
232.0°C (32.0 mmHg)
Authentic
Glass bottle
CH3(CH2)17NH2
500 g
octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
REYJJPSVUYRZGE-UHFFFAOYSA-N
octadecan-1-amine
15793
CHEBI:63866
90%
Primary amine (MW 269):> 98%, Sec. + tert. amine (MW 521): < 2%
Octadecylamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life with long lasting effects.
May be fatal if swallo

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 204-695-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only