missing translation for 'onlineSavingsMsg'
Learn More

Octanohydroxamic Acid 99.0+%, TCI America™

$165.90 - $493.97

Chemical Identifiers

CAS 7377-03-9
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00143918
InChI Key RGUVUPQQFXCJFC-UHFFFAOYSA-N
Synonym Caprylohydroxamic Acid, N-Hydroxyoctanamide
PubChem CID 23846
IUPAC Name N-hydroxyoctanamide
SMILES CCCCCCCC(=O)NO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C12325G
View Documents
TCI America
C12325G
5 g N/A
Only null left
 
C123225G
View Documents
TCI America
C123225G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7377-03-9
159.23
RGUVUPQQFXCJFC-UHFFFAOYSA-N
23846
CCCCCCCC(=O)NO
C8H17NO2
MFCD00143918
Caprylohydroxamic Acid, N-Hydroxyoctanamide
N-hydroxyoctanamide

Specifications

7377-03-9
White
MFCD00143918
Caprylohydroxamic Acid, N-Hydroxyoctanamide
CCCCCCCC(=O)NO
159.23
159.23
Crystalline Powder
78°C
C8H17NO2
5 g
RGUVUPQQFXCJFC-UHFFFAOYSA-N
N-hydroxyoctanamide
23846
≥99.0% (N)
Octanohydroxamic Acid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : RG9900500

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'