missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Octopamine hydrochloride, 99%, Thermo Scientific Chemicals

alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator

$63.18 - $238.27

Chemical Identifiers

CAS 770-05-8
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012881
InChI Key PUMZXCBVHLCWQG-UHFFFAOYNA-N
Synonym octopamine hydrochloride, dl-octopamine hydrochloride, octopamine hcl, epirenor, +/--octopamine hydrochloride, +,--octopamine hcl, 4-2-amino-1-hydroxyethyl phenol hydrochloride, +,--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride
PubChem CID 102484
IUPAC Name 4-(2-amino-1-hydroxyethyl)phenol hydrochloride
SMILES Cl.NCC(O)C1=CC=C(O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6128103
View Documents
Thermo Scientific Chemicals
J6128103
1 g
Each for $63.18
 
AAJ6128106
View Documents
Thermo Scientific Chemicals
J6128106
5 g
Each for $150.85
 
AAJ6128109
View Documents
Thermo Scientific Chemicals
J6128109
10 g
Each for $238.27
 
Description

Description

Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

Solubility
It is soluble in water.

Notes
Handle under argon. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

770-05-8
189.64
PUMZXCBVHLCWQG-UHFFFAOYNA-N
102484
Cl.NCC(O)C1=CC=C(O)C=C1
C8H12ClNO2
MFCD00012881
octopamine hydrochloride, dl-octopamine hydrochloride, octopamine hcl, epirenor, +/--octopamine hydrochloride, +,--octopamine hcl, 4-2-amino-1-hydroxyethyl phenol hydrochloride, +,--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride
4-(2-amino-1-hydroxyethyl)phenol hydrochloride

Specifications

770-05-8
White
MFCD00012881
3915414
octopamine hydrochloride, dl-octopamine hydrochloride, octopamine hcl, epirenor, +/--octopamine hydrochloride, +,--octopamine hcl, 4-2-amino-1-hydroxyethyl phenol hydrochloride, +,--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride
PUMZXCBVHLCWQG-UHFFFAOYNA-N
4-(2-amino-1-hydroxyethyl)phenol hydrochloride
102484
99%
(+/-)-Octopamine hydrochloride
170°C (decomposition)
C8H12ClNO2
1 g
14,6759
It is soluble in water.
Cl.NCC(O)C1=CC=C(O)C=C1
189.64
189.64
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 212-216-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only