missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
$134.31 - $408.23
Chemical Identifiers
CAS | 5466-77-3 |
---|---|
Molecular Formula | C18H26O3 |
Molecular Weight (g/mol) | 290.40 |
MDL Number | MFCD00072582 |
InChI Key | YBGZDTIWKVFICR-UHFFFAOYNA-N |
Synonym | bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate |
PubChem CID | 11044481 |
IUPAC Name | [(2S)-2-ethylhexyl] (E)-3-(4-methoxyphenyl)prop-2-enoate |
SMILES | CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC291160250
|
Thermo Scientific Chemicals
291160250 |
25 g | Glass bottle |
Each for $134.31
|
|
||||
AC291161000
|
Thermo Scientific Chemicals
291161000 |
100 g | Glass bottle |
Each for $408.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5466-77-3 | |
290.40 | |
YBGZDTIWKVFICR-UHFFFAOYNA-N | |
11044481 | |
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1 |
C18H26O3 | |
MFCD00072582 | |
bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate | |
[(2S)-2-ethylhexyl] (E)-3-(4-methoxyphenyl)prop-2-enoate |
Specifications
5466-77-3 | |
830 (at 307 to 308nm in methanol) min. | |
1.0090g/mL | |
Authentic | |
Glass bottle | |
1.5430 to 1.5470 | |
25 g | |
15,6859 | |
YBGZDTIWKVFICR-UHFFFAOYNA-N | |
[(2S)-2-ethylhexyl] (E)-3-(4-methoxyphenyl)prop-2-enoate | |
0.05 to 0.1% BHT | |
290.39 | |
Liquid |
1mg KOH/g max. | |
Colorless to Yellow | |
193°C | |
97.5% min. (GC) | |
C18H26O3 | |
MFCD00072582 | |
1.009 | |
bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate | |
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1 | |
290.40 | |
11044481 | |
98% | |
Octyl 4-methoxycinnamate |
Safety and Handling
EINECSNumber : 226-775-7
RUO – Research Use Only