missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ofloxacin, Thermo Scientific Chemicals

Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity

$72.05 - $259.41

Chemical Identifiers

CAS 82419-36-1
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
MDL Number MFCD00226105
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYSA-N
Synonym Oflocet; Ofloxacine
PubChem CID 4583
ChEBI CHEBI:7731
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6208003
View Documents
Thermo Scientific Chemicals
J6208003
1 g
Each for $72.05
 
AAJ6208006
View Documents
Thermo Scientific Chemicals
J6208006
5 g
Each for $259.41
 
Description

Description

Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

82419-36-1
361.373
GSDSWSVVBLHKDQ-UHFFFAOYSA-N
4583
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
C18H20FN3O4
MFCD00226105
Oflocet; Ofloxacine
CHEBI:7731

Specifications

Ofloxacin
82419-36-1
White to Yellow
C18H20FN3O4
14,6771
Soluble in acetic acid or water; Slightly soluble in methanol
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
4583
361.37
Crystalline Powder
270°C to 275°C
1 g
MFCD00226105
Oflocet; Ofloxacine
GSDSWSVVBLHKDQ-UHFFFAOYSA-N
361.373
CHEBI:7731
Ofloxacin
Safety and Handling

Safety and Handling

RTECSNumber : UU8815500

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only