OHAUS™ Bench Scales Computer Data Transfer Cables
Connects Link Champ General-Purpose Bench Scales to IBM-PC computers via standard RS-232-C.
$412.11 - $412.11
Specifications
Connection Type | RS-232 cable, PC 9 pin |
---|---|
For Use With (Application) | Computer data transfer |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
01-918-26
|
OHAUS™
12106964 |
Models B10S05, B25S05, B50S05, B100S05, B150S05 |
Each for $412.11
|
|
|||||
Description
Permits data collection via computer.
Specifications
RS-232 cable, PC 9 pin | |
Computer data transfer |