OHAUS™ Pull-Spring Scales
Designed to deliver reproducible results time after time, perfect for general weighing, force experiments, action-reaction demonstrations and much more. The Ohaus Demonstration Spring Scales offers a large faceplate for easy viewing.
Supplier:  OHAUS™ 80000041
 View more versions of this product
Catalog No. 021101
Explore available promotions
Description
Description
  • ABS housing with metal face plate, pointer, spring and hook.
  • Displays measurements in Grams, Ounces, Pounds and Newtons.
  • Large faceplate for easy viewing.
  • Weighs in N and hg.
  • Custom internal spring mechanism
Specifications
Specifications
ABS Plastic
18 oz., 500 g
Pull Spring Scale
For general weighing, force experiments, action-re
Analog