OHAUS™ Pull-Spring Scales
Designed to deliver reproducible results time after time, perfect for general weighing, force experiments, action-reaction demonstrations and much more. The Ohaus Demonstration Spring Scales offers a large faceplate for easy viewing.
Supplier:  OHAUS™ 80000044
 View more versions of this product
Catalog No. 0191986
Explore available promotions
Description
Description
  • ABS housing with metal face plate, pointer, spring and hook.
  • Displays measurements in Grams, Ounces
  • Large faceplate for easy viewing.
  • Custom internal spring mechanism
Specifications
Specifications
1.75 in.
13 in.
Stainless Steel
7.62 cm
72 oz., 2 kg
Spring Mechanical Scale
4.44 cm
33 cm
3 in.
For general weighing, force experiments, action-re
Analog