missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Starter pH Pen Meters

Convenient pH testing. OHAUS Starter pH Pen Meters offer a convenient way to measure pH, or the relative acidity or alkalinity of a sample at a given temperature. The pH pen meters can be completely operated with just one hand, freeing up your other hand to handle samples.

$187.82 - $320.13

Specifications

For Use With (Application) Measuring pH, or the relative acidity or alkalinity of asample at a given temperature
Range (pH) 0 to 14
Weight (Metric) 110 g
Weight (English) 0.24 lb.
Battery Type 4 x AG13 1.5V Micro Alkaline
View More Specs

Products 2
Catalog Number Mfr. No. Accuracy (pH) Range (pH) Resolution (pH) Display Type Price Quantity  
Catalog Number Mfr. No. Accuracy (pH) Range (pH) Resolution (pH) Display Type Price Quantity  
S05084
View Documents
OHAUS™
30073970
±0.1 0 to 14 0.1 Dual
Each for $187.82
 
S05085
View Documents
OHAUS™
30073971
0.01 0 to 14 0.01 Single Line
Each for $320.13
 
Description

Description

  • Fast, straightforward results
  • Durable IP67 waterproof design
  • Equipped with wrist strap for security
  • Automatic Shutdown after six minutes of non-use
  • Requires four AG13 1.5V Micro Alkaline Batteries
  • Size/Weight: 185 × 45 × 38mm, 110g
  • Includes buffer powder pack (ST20)
Specifications

Specifications

Measuring pH, or the relative acidity or alkalinity of asample at a given temperature
110 g
4 x AG13 1.5V Micro Alkaline
0 to 14
0.24 lb.
Portable pH Meter
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'