missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
OHAUS™ Triple Beam Balances
Accurate, rugged and dependable. OHAUS Triple Beam Balances have stainless-steel weighing pans and bearing covers which prevent rust and corrosion.
$421.46 - $421.46
Specifications
Diameter (English) Outer Pan | 6 in. |
---|---|
Diameter (Metric) Outer Pan | 15.2 cm |
Depth (English) | 9 in. |
Depth (Metric) | 22.86 cm |
Height (English) | 12 in. |
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Catalog Number | Mfr. No. | Model | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Material | Price | Quantity | ||||
S40020
|
OHAUS™
750SO |
750-S0 | Stainless Steel |
Each for $421.46
|
|
||||
Description
- Precision-cast poises and deep notches in beams for accurate values every time
- Precision-ground knife edges and self-aligning agate bearings for maximum sensitivity and reproducibility
- Accurate with quick response time
- Magnetic damping minimizes oscillation
- Heavy zinc base won't tip easily like cast aluminum
- Capacity: 610g (2610g with attachment weights)
- Readability: 0.1g
- Front Beam: 10 × 0.1g
- Middle Beam: 500 × 100g
- Rear Beam: 100 × 10g
- Vernier Dial (Dial-O-Gram™ Model only): 10 × 0.1g
- Pan size: 6 in. dia. (21.2cm)
- Dimensions: 18L × 12W × 9 in.H (45.7 × 30.5 × 22.3cm)
Dial-O-Gram model includes advanced features
- Direct-reading vernier dial for faster weighing results
- Tare beam allows containers up to 225g to be balanced out
- Add rod and clamp accessory (S40022B) to perform specific gravity determinations
Specific Gravity Rod and Clamp
- Suspends balance above base of table to measure specific gravity.
Poly Scoop and Counterweight
- Ideal for retaining all of the object being weighed
- Counterweight returns balance to zero when nothing but the scoop is on the platform
- Stainless-steel scoop and counterweight feature heavy-duty construction for long wear
Note
S40028 features a plastic container
Warranty
Five years
Specifications
6 in. | |
9 in. | |
12 in. | |
18 in. | |
5 Years |
15.2 cm | |
22.86 cm | |
30.48 cm | |
45.72 cm | |
1.34 lb., 0.61 kg |