OHAUS™ Valor™ 3000 Compact Food Scales
Stainless-steel design in a high-performance, easy-to-use, portable scale
Supplier:  OHAUS™ 83998178
 View more versions of this product
Catalog No. 02-115-331
Explore available promotions
Description
Description
  • Software for weighing, percent weighing and specific gravity determination
  • All stainless-steel construction
  • Compact space-saving footprint
  • AC adapter (supplied) or 4 C batteries
  • Adjustable levelling feet and bubble (only for XW models)
  • Load cell lock, user adjustable filtering and optional food accessories
  • LCD with white LED backlight
Certifications
NSF Certified
Compliance
Supports HACCP compliance; washable models available.
Specifications
Specifications
3 Mechanical switch
4 C
ISO 9001:2008, NSF/ANSI 169-2005, USDA/AMS, HACCP, CE, AS/NZS4251.1 Emission/AS/NZS4252.1 Immunity, FCC Class B Part 15 operation, Industry Canada Class A, cULus AC power pack, Directive 2002/96/EC, NTEP CoC No. 08-008
LCD
Gram, kilogram, ounce, pound, fluid ounce
Stainless Steel
<3 sec.
0°C to 40°C
8.5 x 7.3 x 3 in. (21.8 x 18.6 x 7.7 cm)
21.8 cm
18.6 cm
7.7 cm
1.2 kg
100/240 V
50 Hr.
Digital external span calibration
400 g
0.01 g
AC adapter, adjustable leveling feet and bubble, load cell lock
V31XH402
32°F to 104°F
Valor™ 3000 Compact Food Scale
8.5 in.
7.3 in.
3 in.
2.6 lb.
100/240 V 50/60 Hz
50/60 Hz