Troemner™ OIML Weight Cases
Polycarbonate Case for OIML Precision Weight
Supplier:  Troemner™ 80864279
 View more versions of this product
Catalog No. 02-215-562
Explore available promotions
Description
Description
  • Polycarbonate Case for OIML Precision Weight has a comfortable handle, a recessed grip for removing the lid, side latches for holding the lid in place, and a color-coded insert.
  • Urethane bumpers, both inside the lid and on the bottom of the case, gently hold the weight in place.
Specifications
Specifications
Individual OIML Weight Case
Polycarbonate
1g OIML weight
Each