Troemner™ OIML Weight Cases
Polycarbonate Case for OIML Precision Weight
Supplier: Troemner™ 80864288
Description
- Polycarbonate Case for OIML Precision Weight has a comfortable handle, a recessed grip for removing the lid, side latches for holding the lid in place, and a color-coded insert.
- Urethane bumpers, both inside the lid and on the bottom of the case, gently hold the weight in place.
Specifications
Individual OIML Weight Case | |
Polycarbonate | |
200g OIML weight | |
Each |