Learn More
Oleic acid, 99%, Thermo Scientific Chemicals
$119.86 - $1165.93
Chemical Identifiers
CAS | 112-80-1 |
---|---|
Molecular Formula | C18H34O2 |
Molecular Weight (g/mol) | 282.47 |
MDL Number | MFCD00064242 |
InChI Key | ZQPPMHVWECSIRJ-MDZDMXLPSA-N |
Synonym | oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid |
PubChem CID | 445639 |
ChEBI | CHEBI:16196 |
IUPAC Name | (Z)-octadec-9-enoic acid |
SMILES | CCCCCCCC\C=C\CCCCCCCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3199706
|
Thermo Scientific Chemicals
03199706 |
5 g |
Each for $119.86
|
|
|||||
AA3199714
|
Thermo Scientific Chemicals
03199714 |
25 g |
Each for $398.25
|
|
|||||
AA3199722
|
Thermo Scientific Chemicals
03199722 |
100 g |
Each for $1,165.93
|
|
|||||
Description
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in the rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in the oil and gas industry. It finds application as an excipient in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-80-1 | |
282.47 | |
ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
445639 | |
(Z)-octadec-9-enoic acid |
C18H34O2 | |
MFCD00064242 | |
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid | |
CHEBI:16196 | |
CCCCCCCC\C=C\CCCCCCCC(O)=O |
Specifications
112-80-1 | |
Colorless to Yellow | |
286°C (100 mmHg) | |
1.461 | |
5 g | |
Air Sensitive | |
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid | |
ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
(Z)-octadec-9-enoic acid | |
445639 | |
282.47 | |
Liquid |
13°C to 14°C | |
0.887 g/mL | |
C18H34O2 | |
MFCD00064242 | |
1726542 | |
146828 | |
Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide. | |
CCCCCCCC\C=C\CCCCCCCC(O)=O | |
282.47 | |
CHEBI:16196 | |
99% | |
Oleic acid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Warning
EINECSNumber : 204-007-1
RTECSNumber : RG2275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only