missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oleic acid, tech. 90%, Thermo Scientific Chemicals

$96.62 - $241.89

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA16663AU
View Documents
Thermo Scientific Chemicals
A16663AU
1000 mL
Each for $96.62
 
AAA166630J
View Documents
Thermo Scientific Chemicals
A166630J
5000 mL
Each for $241.89
 
Description

Description

Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in oil and gas industry. It finds application as an excipient in pharmaceuticals.

Solubility
Miscible with ethanol, ether, acetone, chloroform, dimethyl formamide and dimethyl sulfoxide.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and aluminum.
Specifications

Chemical Identifiers

112-80-1
282.47
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
445639
CCCCCCCC\C=C\CCCCCCCC(O)=O
C18H34O2
MFCD00064242
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
CHEBI:16196

Specifications

112-80-1
0.887 g/mL
90%
1.461
1000 mL
Air sensitive
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
(9E)-octadec-9-enoic acid
445639
282.47
Technical
13°C to 14°C
286°C (100 mmHg)
C18H34O2
MFCD00064242
1726542
14,6828
Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide.
CCCCCCCC\C=C\CCCCCCCC(O)=O
282.47
CHEBI:16196
90%
Oleic Acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319

GHS P Statement
P280-P264-P305+P351+P338-P362-P321-P332+P313-P337+P313-P302+P352

Warning

EINECSNumber : 204-007-1

RTECSNumber : RG2275000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only