missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oleoyl Chloride 80.0+%, TCI America™

Supplier:  TCI America O005325G

 View more versions of this product

Catalog No. O005325G


Explore available promotions

Description

Description

Specifications

Specifications

Oleoyl Chloride
Yellow
C18H33ClO
25 g
elaidoylchloride, elaidoyl chloride, 9-octadecenoyl chloride, oleoylchloride, elaidic acid chloride, trans-9-octadecenoyl chloride, e-octadec-9-enoyl chloride, 9e-octadec-9-enoyl chloride
CCCCCCCC\C=C/CCCCCCCC(Cl)=O
300.91
300.91
Liquid
112-77-6
200°C
MFCD00134332
3265
MLQBTMWHIOYKKC-KTKRTIGZSA-N
(9Z)-octadec-9-enoyl chloride
5702750
≥80.0% (GC)

Chemical Identifiers

112-77-6
300.91
MLQBTMWHIOYKKC-KTKRTIGZSA-N
5702750
CCCCCCCC\C=C/CCCCCCCC(Cl)=O
C18H33ClO
MFCD00134332
elaidoylchloride, elaidoyl chloride, 9-octadecenoyl chloride, oleoylchloride, elaidic acid chloride, trans-9-octadecenoyl chloride, e-octadec-9-enoyl chloride, 9e-octadec-9-enoyl chloride
(9Z)-octadec-9-enoyl chloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3552

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions