missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oleyl Alcohol 60.0+%, TCI America™
Supplier: TCI America O0058500ML
Specifications
Oleyl Alcohol | |
Yellow | |
C18H36O | |
500 mL | |
ALSTYHKOOCGGFT-KTKRTIGZSA-N | |
(9Z)-octadec-9-en-1-ol | |
5284499 | |
268.49 | |
Liquid |
143-28-2 | |
207°C | |
MFCD00002993 | |
oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol | |
CCCCCCCC\C=C/CCCCCCCCO | |
268.49 | |
CHEBI:73504 | |
≥60.0% (GC) |
Chemical Identifiers
143-28-2 | |
268.49 | |
ALSTYHKOOCGGFT-KTKRTIGZSA-N | |
5284499 | |
(9Z)-octadec-9-en-1-ol |
C18H36O | |
MFCD00002993 | |
oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol | |
CHEBI:73504 | |
CCCCCCCC\C=C/CCCCCCCCO |
Safety and Handling
EINECSNumber : (2)-0258
RTECSNumber : RG4120000
TSCA : Yes