missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Omeprazole, 98%, Thermo Scientific Chemicals

A proton pump inhibitor

Supplier:  Thermo Scientific Chemicals J6286003

 View more versions of this product

Catalog No. AAJ6286003


Explore available promotions

Description

Description

Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.

Solubility
Soluble in water (0.5 mg/ml), DMSO (25 mg/ml), and ethanol (4.5 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications

Specifications

Omeprazole
158°C to 159°C
C17H19N3O3S
1 g
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
SUBDBMMJDZJVOS-UHFFFAOYSA-N
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
4594
345.42
Powder
73590-58-6
Brown to White
MFCD00083192
14,6845
Soluble in water (0.5mg/ml),DMSO (25mg/ml),and ethanol (4.5mg/ml).
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
345.417
CHEBI:77260
98%

Chemical Identifiers

73590-58-6
345.417
SUBDBMMJDZJVOS-UHFFFAOYSA-N
4594
6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
C17H19N3O3S
MFCD00083192
omeprazole, losec, prilosec, antra, esomeprazole, omeprazon, audazol, omapren, omepral, parizac
CHEBI:77260
CC1=CN=C(C(=C1OC)C)CS(=O)C2=NC3=C(N2)C=C(C=C3)OC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : DD9087000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only