Promega ONE-Glo™ EX Luciferase Assay System DFS Item
ONE-Glo™ EX Luciferase Assay System provides both the high sensitivity and long-lived luminescence required to batch-process multiple 96-, 384- or 1,536-well plates. The approximately 2-hour signal half-life provides greater flexibility in assay design.
Supplier:  Promega E8130
 View more versions of this product
Catalog No. PRE8130
Explore available promotions
Description
Description
  • Highly Sensitive Assay with Long-Lived Luminescence
  • Bright, stable signal that can be measured for hours
  • Stable reagent can be stored at room temperature or 4°C
  • Reduced sensitivity to culture media, phenol red and luciferase inhibitors
Specifications
Specifications
10 x 10 mL
Product Suggestions
Product Suggestions