Learn More
Dunlop™ Onguard™ Hazmax™ EZ-Fit™ Boots
Designed for easy donning and doffing while wearing chemical protective suit
$263.21 - $279.00
Specifications
For Use With (Application) | For situation involving dangerous chemicals, gases and waste |
---|---|
Height (English) | 11 in. |
Height (Metric) | 28 cm |
Alert:
Caution! Do not use for fire or electrical hazard protection.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
29-635-4055
|
Dunlop™
87015L |
Large |
N/A
|
|
|||||
29-635-4053
|
Dunlop™
87015S |
Small |
N/A
|
|
|||||
29-635-4056
|
Dunlop™
87015XL |
X-Large |
N/A
|
|
|||||
Description
- One-piece injection molded with no buttons or elastic bands to break for easier decontamination
- Large kick-off lug for easy removal
- Wider across the instep for easy entry
- Nonslip EZ decon outsole for easier decontamination and cleanup
- Steel toe, steel shank, and steel midsole for complete protection
- Height: 11 in. (28cm)
Order Info
Order normal shoe size–designed to accommodate a fully encapsulated suit. Men's full sizes: Small: 6-8, Medium: 9-10, Large: 11-12, X-Large: 13-15.
Certifications
Certified by S.E.I. to meet the requirements of the NFPA 1991, 2005 Edition Standard on Vapor-Protective ensembles for Hazardous Materials Emergencies.
Compliance
Meets ASTM™ F2413-11; CSA Z195 requirements. NFPA 1991 Standard on Vapor-Protective Ensembles for Hazardous Materials Emergencies and CBRN Terrorism Incidents, 2016.
Specifications
For situation involving dangerous chemicals, gases and waste | |
11 in. | |
28 cm |