missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Open Head Steel Drum
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
Supplier: Youngstown Barrel & Drum 445L
Description
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.
- Salvage drum designed to overpack and ship damaged and/or leaking drums
- Also qualify for direct containment of hazardous solids
- Freight Class 50
Specifications
Lined Drum | |
18 Gauge | |
Black, White | |
Steel | |
Open | |
28 in. | |
18.25 in. | |
35 lb. |
UN 1A2/Y1.5/150/X250/S-DOT/UN | |
114 L | |
For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
50 | |
Ea. | |
71.12 cm | |
46.35 cm | |
15.87 Kg |