missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Youngstown Barrel & Drum Open Head Steel Drum
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
Supplier: Youngstown Barrel & Drum YBD220
Description
Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.
- Salvage drum designed to overpack and ship damaged and/or leaking drums
- Also qualify for direct containment of hazardous solids
- Freight Class 50
Specifications
Unlined Drum | |
Bung | |
208.19 L | |
For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
50 | |
Ea. | |
84.45 cm | |
57.15 cm | |
22.22 Kg |
UN 1A2/Y1.6/150/X400/S-DOT/UN | |
18/16 Gauge | |
Black, White | |
Steel | |
Open | |
33.25 in. | |
22.5 in. | |
49 lb. |