MilliporeSigma™ Supelco™ OPTI-SEAL Pump Seal
OPTI-SEAL pump seals are made from an inert, ultra-high molecular weight polyethylene material (UHMW-PE). Relative to conventional polytetrafluoroethylene (PTFE) seals, these seals show less particle shedding. This reduces the potential for clogged frits, resulting in the prevention of unscheduled system downtime.
$133.90 - $140.98
Specifications
For Use With (Application) Liquid Chromatography
View More Specs
Products 2
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Description For Use With (Equipment) Price Quantity  
111004828
View Documents
MilliporeSigma™ Supelco™
59407
Mfg Part Number: Hitachi 655-1080 655, L6000, L6200, L6200A
Each for $140.98
111004742
View Documents
MilliporeSigma™ Supelco™
59492
Mfg Part Number: LDC/Milton Roy 206234 All Analytical Pumps
Each for $133.90
Description
Description
These seals are recommended for typical reversed phase mobile phases (aqueous to moderate levels of organic). They are not recommended for long-term use with highly organic mobile phases, such as typical normal phase solvents.
Specifications
Specifications
Liquid Chromatography
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'