Optima™ LC/MS Solvents Starter Kit, Includes 1L Acetonitrile, 1L Methanol, 1L Ipa And 1L Water Optima™ LC/MS Grade
Starter kit with reagents and amber glass bottle for development of the mobile phase for LC/MS
Supplier:  Thermo Fisher Scientific LCMSKIT
Catalog No. LCMSKIT
Explore available promotions
Description
Description
  • The starter kit includes a 1L amber glass bottle for each of the following reagents:
    • A955-1, Optima LC/MS Acetonitrile
    • A456-1. Optima LC/MS Methanol
    • A461-1, Optima LC/MS IPA
    • W6-1, Optima LC/MS Water
  • 1L empty amber glass bottle is included to mix reagents for the mobile phase
Specifications
Specifications
1 Box
Creating Mobile Phase
LC/MS Solvents Starter Kit