OPTIZYME™ 10X Buffer 1, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80841
Catalog No. BP8084-1
Explore available promotions
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.1mg/mL BSA
Specifications
Specifications
10mM Tris HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.1mg/mL BSA
5 x 1 mL
Liquid
-20°C
Buffer 1