OPTIZYME™ 10X Buffer 4, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80871
Catalog No. BP8087-1
Explore available promotions
Description
Description
1X Buffer Composition:
33mM Tris-acetate (pH 7.9 at 37°C), 10mM magnesium acetate, 66mM potassium acetate, 0.1mg/mL BSA
Specifications
Specifications
33mM Tris Acetate (pH 7.9 at 37°C), 10mM Magnesium Acetate, 66mM Potassium Acetate, 0.1mg/mL BSA
5 x 1 mL
Liquid
-20°C
Buffer 4