OPTIZYME™ 10X Buffer AarI, ScaI, PasI, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80961
Catalog No. BP8096-1
Explore available promotions
Description
Description
1X Buffer Composition:
10mM Bis-Tris Propane-HCl (pH 6.5 at 37°C), 10mM MgCl2, 100mM KCl, 0.1mg/mL BSA
Specifications
Specifications
Bis-tris Propane-HCl 10mM (pH 6.5 at 37°C), 10mM MgCl2, 100mM KCl, 0.1mg/mL BSA
5 x 1 mL
Liquid
-20°C
Buffer AarI, ScaI, PasI