OPTIZYME™ 10X Buffer Cfr9I, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80931
Catalog No. BP8093-1
Explore available promotions
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 7.2 at 37°C), 5mM MgCl2, 200mM sodium glutamate and 0.1mg/mL BSA
Specifications
Specifications
10mM Tris HCl (pH 7.2 at 37°C), 5mM MgCl2, 200mM Sodium Glutamate, 0.1mg/mL BSA
1 mL
Liquid
Keep container tightly closed
Buffer Cfr9I