OPTIZYME™ 10X Buffer KpnI, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80941
Catalog No. BP8094-1
Explore available promotions
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.02% Triton X-100 and 0.1mg/mL BSA
Specifications
Specifications
10mM Tris HCl (pH 7.5 at 37°C), 10mM MgCl2, Triton™ X 100 0.02%, 0.1mg/mL BSA
1 mL
Liquid
-20°C
10X Buffer KpnI