OPTIZYME™ 10X Buffer Sau3AI, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80911
Catalog No. BP8091-1
Explore available promotions
Description
Description
1X Buffer Composition:
33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate, 0.02% Triton X-100 and 0.1mg/mL BSA
Specifications
Specifications
33mM Tris Acetate (pH 7.9 at 37°C), 10mM Magnesium Acetate, 66mM Potassium Acetate, 0.1mg/mL BSA, 0.02% Triton™ X-100
1 mL
Liquid
-20°C
Sau3AI