OPTIZYME™ 10X Buffer TaqI, Fisher BioReagents™
Supplier:  Thermo Fisher Scientific BP80921
Catalog No. BP8092-1
Explore available promotions
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 8.0 at 37°C), 5mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
Specifications
Specifications
10mM Tris HCl (pH 8 at 37°C), 5mM MgCl2, 100mM NaCl, 0.1mg/mL BSA
1 mL
Liquid
-20°C
Taql