missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OPTIZYME™ BglII, Fisher BioReagents™

$1074.02 - $1074.02

Specifications

Concentration 10 U/μL
Components 25 to 50% Water, >50% Glycerin, 1 to 2.5% Sodium Chloride
Incubator Temperature 37°C
pH 7.4
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BglII
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BP8014-5
View Documents
Thermo Fisher Scientific
BP80145
2500 U
Each for $1,074.02
 
Description

Description

5'...A^G A T C T...3'
3'...T C T A G^A...5'

Supplied with: 10X OPTIZYME Buffer 3
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 3: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: 20 - 50%
  • Buffer 3: 100%
  • Buffer 4: 0 - 20%
  • Buffer 5: 50 - 100%
Storage Buffer:
  • 10mM Tris-HCl (pH 8.2 at 25°C), 200 mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after a 50-fold over-digestion with BglII.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of BglII is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Compatible Ends: BamHI, BclI, Bsp143I, MboI, PsuI
  • Methylation Effects:
  • Dam: Completely overlaps - no effect
    • AGm6ATCT
  • (No effect on cleavage)
  • Dcm: Never overlaps - no effect
  • CpG: Never overlaps - no effect
  • EcoKI: Never overlaps - no effect
  • EcoBI: May overlap - cleavage impaired
    • 5'...TGm6AGATCT(N)3 TGCT...3'
  • 3'...AC TCTAGA(N)3m6ACGA...5'
  • (Cleavage is impaired)
  • Specifications

    Specifications

    10 U/μL
    37°C
    >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BglII
    Keep container tightly closed
    BglII
    25 to 50% Water, >50% Glycerin, 1 to 2.5% Sodium Chloride
    7.4
    10mM Tris HCl (pH 8.2 at 25°C), 200mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
    A.GATCT
    Liquid
    Videos
    SDS
    missing translation for 'documents'

    missing translation for 'documents'