missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OPTIZYME™ EcoRI, Fisher BioReagents™

Supplier:  Thermo Fisher Scientific BP80541

 View more versions of this product

Catalog No. BP8054-1


Explore available promotions

Description

Description

5'...G^A A T T C...3'
3'...C T T A A^G...5'

Conditions for 100% Activity:

  • 1X OPTIZYME Buffer EcoRI: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl, 0.02% Triton X-100 and 0.1mg/ml BSA.
  • Incubate at 37°C.
Supplied with: 10X OPTIZYME EcoRI Buffer
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: No reaction
  • Buffer 3: 100%
  • Buffer 4: No reaction
  • Buffer 5: 100%™
™star activity occurs at greater than 5-fold overdigestion (5 units X 1 hour)
Storage Buffer:
10mM potassium phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/ml BSA, 0.15% Triton X-100 and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with EcoRI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of EcoRI is required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Note: Low salt concentration, large excess of the enzyme, pH>8.0, or the replacement of mg2+ by Mn2+ may result in star activity.
Compatible Ends: XapI, MunI, TasI.
Methylation Effects:
Dam: never overlaps - no effect.
Dcm: never overlaps - no effect.
CpG: may overlap - cleavage impaired.
  • 5'...m5C GAATTm5C G...3'
  • 3'... Gm5CTTAA Gm5...5'
  • EcoKI: never overlaps - no effect.
  • EcoBI: may overlap - no effect.
    • 5'...TGm6AATTC(N)4TGCT...3'
  • 3'...AC TTAAG(N)4m6ACGA...5'
  • Specifications

    Specifications

    50 U/μL
    37°C
    >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRI
    Keep container tightly closed
    G.AATTC
    Liquid
    25 to 50% Water, >50% Glycerin, 2.5 to10% Sodium Chloride
    7.4
    10mM Potassium Phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
    25,000 U
    EcoRI
    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions