missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OPTIZYME™ NheI, Fisher BioReagents™

Supplier:  Thermo Fisher Scientific BP80191

Catalog No. BP8019-1


Explore available promotions

Description

Description

5'...G^C T A G C...3'
3'...C G A T C^G...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 100%
  • Buffer 2: 20 - 50%
  • Buffer 3: 0 - 20%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 8.0 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with NheI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of NheI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Notes:
    • Low salt, high glycerol (>5%) concentrations, pH>8.0 or a large excess of enzyme may result in star activity.
    • NheI is inhibited by salt concentrations above 100mM.
    • Supercoiled plasmids may require up to 10-fold more NheI for complete digestion than linear DNAs (e.g. 10 units are required to cleave 1μg of pBR322 DNA).
    Compatible Ends: XmaJI, BcuI, Eco130I, XbaI
    Methylation Effects:
    Dam: Never overlaps - no effect
    Dcm: Never overlaps - no effect
    CpG: May overlap - cleavage impaired
    • 5'...GCTAGm5C G...3'
  • 3'...CGATC Gm5C...5'
  • (Cleavage is impaired)
    • 5'...m5C GCTAGm5C G...3'
  • 3'... Gm5CGATC Gm5C...5'
  • (Cleavage is blocked)
  • EcoKI: Never overlaps - no effect
  • EcoBI: Never overlaps - no effect
  • Specifications

    Specifications

    10 U/μL
    7.4
    10mM Tris HCl (pH 8 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    500 U
    NheI
    25 to 50% Water, >50% Glycerin
    >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Nhe I
    Keep container tightly closed
    G.CTAGC
    Liquid
    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions