missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
OPTIZYME™ NsiI (Mph1103I) Fisher BioReagents™
Supplier: Fisher BioReagents BP80581
Description
5'...A T G C A^T...3'
3'...T^A C G T A...5'
Supplied with: 10X OPTIZYME Buffer 5
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 0 – 20%
- Buffer 2: 50 - 100%
- Buffer 3: 20 - 50%
- Buffer 4: 50 - 100%
- Buffer 5: 100%
- 10mM Tris-HCl (pH 7.4 at 25°C), 200mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
Specifications
10 U/μL | |
7.4 | |
10mM Tris HCl (pH 7.4 at 25°C), 200mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol | |
1000 U | |
NsiI |
25 to 50% Water, >50% Glycerin, 1 to 2.5% Potassium Chloride | |
>90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Not I | |
Keep container tightly closed | |
ATGCA.T | |
Liquid |