missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Supelco™ ORBO™ 353 HBr on Petroleum Charcoal (20/40), 100/50 mg
Solvent desorption tube designed for gas and vapor sampling.
Supplier: MilliporeSigma™ Supelco™ 20044
Description
- Contains two beds of the same selective adsorbent (Bed A: 100 mg, Bed B: 50 mg) separated by glass wool or foam
- Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed
- Matrix: HBr on Petroleum Charcoal
Specifications
HBr on Petroleum Charcoal | |
Gas and Vapor Sampling | |
Solvent Desorption Tube | |
6 mm |
20 to 40 Mesh | |
25 | |
75 mm |
ORBO is a trademark of Sigma-Aldrich Co. LLC