missing translation for 'onlineSavingsMsg'
Learn More

Orcinol, 98%

CAS: 504-15-4 | C7H8O2 | 124.14 g/mol

$105.56 - $303.81

Chemical Identifiers

CAS 504-15-4
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002291
InChI Key OIPPWFOQEKKFEE-UHFFFAOYSA-N
Synonym orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
PubChem CID 10436
ChEBI CHEBI:16536
IUPAC Name 5-methylbenzene-1,3-diol
SMILES CC1=CC(O)=CC(O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC416580050
SDS View Documents
Thermo Scientific Chemicals
416580050
5 g Glass bottle
Each for $105.56
Only null left
 
AC416580250
SDS View Documents
Thermo Scientific Chemicals
416580250
25 g Glass bottle
Each for $303.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

504-15-4
124.14
OIPPWFOQEKKFEE-UHFFFAOYSA-N
10436
5-methylbenzene-1,3-diol
C7H8O2
MFCD00002291
orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
CHEBI:16536
CC1=CC(O)=CC(O)=C1

Specifications

504-15-4
Brown-Pink to Gray-Pink
159°C
97.5% min. (GC)
C7H8O2
5 g
orcinol, 3,5-dihydroxytoluene, 5-methylresorcinol, 1,3-dihydroxy-5-methylbenzene, orcin, 5-methyl-1,3-benzenediol, 5-methylresorcin, 3-hydroxy-5-methylphenol, 3,5-toluenediol, 1,3-benzenediol, 5-methyl
CC1=CC(O)=CC(O)=C1
124.14
CHEBI:16536
98%
Orcinol
105.0°C to 110.0°C
290.0°C
Authentic
Glass bottle
MFCD00002291
15,6959
OIPPWFOQEKKFEE-UHFFFAOYSA-N
5-methylbenzene-1,3-diol
10436
124.14
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 207-984-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only