missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Orion™ 8102BNUWP ROSS Ultra™ pH Electrode
Conduct precise pH determinations with this general purpose, top performance glass body combination electrode with waterproof BNC connector.
Supplier: Thermo Scientific™ ORI8102BNUWP
Description
Conduct precise pH determinations with the Thermo Scientific™ Orion™ 8102BNUWP ROSS Ultra™ pH Electrode, a general purpose, top performance glass body combination electrode with waterproof BNC connector. The pH response is independent of temperature changes and can be used on any pH meter with a BNC connection.
Includes:
- 60ml bottle of Orion ROSS electrode fill solution, 810007
Requires:
- pH buffers for calibration
- Beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Compatible with:
Waterproof BNC connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.
Recommended for:
- Q.C. and research applications
Specifications
Glass body combination electrode | |
Ceramic | |
0 to 14 | |
pH Electrode | |
3M KCl | |
ROSS | |
32°F to 212°F | |
1 m |
WPBNC | |
Glass Body | |
0°C to 100°C | |
<0.002pH/day | |
60ml bottle of Orion ROSS electrode fill solution | |
0.01 | |
39.4 in. |