Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories
Ensure your turbidity meter has the accessories it needs, including turbidity vials, primary turbidity standards, Formazin turbidity standards, and more.
$116.76 - $1387.56
Products 4
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
13-642-051
View Documents
Thermo Scientific™
AC45ST
Primary Standards for AQ4500, SDVB for 0, 1.0, 10, 100, 1000NTU
Pack of 5 Each for $1,387.56
13-650-110
View Documents
Thermo Scientific™
AC45S1
Silicon Oil and Cloth Kit
Each for $116.76
13-650-122
View Documents
Thermo Scientific™
AC2T24
Replacement Turbidity Vials
Pack of 12 Each for $670.94
13-642-184
View Documents
Thermo Scientific™
AQ4CBL
RS232 cable for method uploads and data downloads in AQ4000/AQ4500
Each for $455.89
Description
Description
Ensure your Thermo Scientific™ Orion™ AQUAfast™ AQ4500 Turbidity Meter has the accessories it needs with our selection of Orion™ AQUAfast™ AQ4500 Turbidity Meter Accessories that include turbidity vials, primary turbidity standards, Formazin turbidity standards, and more. Our Orion AQUAfast AQ4500 turbidity meter has advanced features, including dual light sources and drinking water / wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols; use the White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
Accessories for the Orion AQUAfast turbidity meter include primary standards, Formazin turbidity standard, silicon oil and cloth kit, and replacement turbidity vials.
Warranty: Out of box
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.