Thermo Scientific™ Orion™ Electrode Adapter Cables
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
$243.41 - $243.41
Products 1
Catalog Number Mfr. No. Connector Types Meter Input Price Quantity  
Catalog Number Mfr. No. Connector Types Meter Input Price Quantity  
13-641-49
View Documents
Thermo Scientific™
91CBNC
Screw Cap BNC
Each for $243.41
Description
Description
Convert an electrode with one connector to the required connector of another meter by using the Thermo Scientific™ Orion™ Electrode Adapter Cables.
  • For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2mm reference pins or just a stripped lead inputs
  • Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors
Warranty: Out of Box
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'