Learn More
Thermo Scientific™ Orion™ pH Buffer Bottles
Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.
$62.59 - $320.26
Specifications
Chemical Name or Material | pH Buffer, bottles |
---|---|
For Use With (Equipment) | pH Meter and Electrode |
Physical Form | Liquid |
Traceability to NIST | Yes |
Catalog Number | Mfr. No. | pH | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | pH | Quantity | Price | Quantity | ||||
13-643-002
|
Thermo Scientific™
ORI910168 |
1.68 ± 0.01 pH | 475 mL |
Each for $96.68
|
|
||||
13-643-007
|
Thermo Scientific™
9116860 |
1.68 ± 0.01 pH | 5 x 60 mL |
Pack of 5 Each for $199.91
|
|
||||
13-079-319
|
Thermo Scientific™
910110 |
10.01 ± 0.02 pH | 475 mL |
Each for $89.65
|
|
||||
09-357-68
|
Thermo Scientific™
ORI911060 |
10.01 ± 0.02 pH | 5 x 60 mL |
Each for $122.35
|
|
||||
13-643-001
|
Thermo Scientific™
910112 |
12.46 ± 0.03 pH | 475 mL |
Each for $104.79
|
|
||||
13-643-005
|
Thermo Scientific™
911260WA |
12.46 ± 0.03 pH | 5 x 60 mL |
Pack of 5 Each for $320.26
|
|
||||
13-301-133
|
Thermo Scientific™
910104 |
4.01 ± 0.01 pH | 475 mL |
Each for $93.76
|
|
||||
09-357-69
|
Thermo Scientific™
910460 |
4.01 ± 0.01 pH | 5 x 60 mL |
Each for $127.98
|
|
||||
13-640-8
|
Thermo Scientific™
910105 |
5.00 ± 0.02 pH | 475 mL |
Each for $62.59
|
|
||||
13-643-003
|
Thermo Scientific™
910686 |
6.86 ± 0.01 pH | 475 mL |
Each for $117.12
|
|
||||
13-643-006
|
Thermo Scientific™
916860 |
6.86 ± 0.01 pH | 5 x 60 mL |
Pack of 5 Each for $210.44
|
|
||||
13-641-857
|
Thermo Scientific™
910107 |
7.00 ± 0.01 pH | 475 mL |
Each for $93.76
|
|
||||
09-357-70
|
Thermo Scientific™
910760 |
7.00 ± 0.01 pH | 5 x 60 mL |
Each for $127.98
|
|
||||
13-643-004
|
Thermo Scientific™
910918 |
9.18 ± 0.01 pH | 475 mL |
Each for $96.24
|
|
||||
13-643-008
|
Thermo Scientific™
9191860 |
9.18 ± 0.01 pH | 5 x 60 mL |
Pack of 5 Each for $224.76
|
|
||||
Description
Use Thermo Scientific™ Orion™ pH buffer bottles to perform accurate, reproducible pH calibrations on your pH meter and pH electrode measurement system. Orion pH buffers are available in 475mL bottles for everyday use and laboratory and bench pH analysis and available in 60 mL bottle 5 packs for occasional use and portable and field pH analysis. Using high-quality pH buffers to calibrate your pH system is the best way to ensure your pH measurements are correct and repeatable.
Whether you are in the lab or in the field, you can be confident that Orion solutions will deliver accurate and repeatable measurements. Orion solutions are produced in controlled batches using ultrapure water and undergo rigorous quality testing throughout the process to prevent contamination. Our strict quality procedures verify correct bottle labeling and volume, so each batch meets specific lot release criteria. Bottles are sealed and torque-tested to ensure bottle caps are secure and solutions remain free of contamination. Rigorous lifetime testing ensures sealed Orion solutions remain within published specifications for their entire shelf life and do not degrade over time.
- Orion pH buffers in 475mL bottles are ideal for everyday use and laboratory and bench pH analysis.
- Orion pH buffers in 60 mL bottle 5 packs are designed for occasional use and portable and field pH analysis.
- Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
- Orion pH buffers are color-coded for easy identification: pH 4 buffer is pink, pH 5 buffer is orange, pH 7 buffer is yellow and pH 10 buffer is blue.
Specifications
pH Buffer, bottles | |
pH Meter and Electrode | |
Liquid | |
Yes |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.