Learn More
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
$150.38 - $228.53
Specifications
Chemical Name or Material | pH Buffer, individual use pouches |
---|---|
Boiling Point | 100°C |
For Use With (Application) | pH calibration |
For Use With (Equipment) | pH Meter and Electrode, single-use |
Physical Form | Liquid |
Catalog Number | Mfr. No. | pH | Packaging | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | pH | Packaging | Quantity | Price | Quantity | |||
13-641-270
|
Thermo Scientific™
910410WA |
4.01 | Pouch | 10 x 15 mL |
Pack of 10 Each for $150.38
|
|
|||
13-641-274
|
Thermo Scientific™
ORI910425 |
4.01 | Pouch | 25 x 15 mL |
Pack of 25 Each for $228.53
|
|
|||
13-641-271
|
Thermo Scientific™
910710 |
7 | Pouch | 10 x 15 mL |
Pack of 10 Each for $150.38
|
|
|||
13-641-275
|
Thermo Scientific™
910725 |
7 | Pouch | 25 x 15 mL |
Pack of 25 Each for $228.53
|
|
|||
13-641-272
|
Thermo Scientific™
911010 |
10.01 | Pouch | 10 x 15 mL |
Pack of 10 Each for $150.38
|
|
|||
13-641-276
|
Thermo Scientific™
ORI911025-WA |
10.01 | Pouch | 25 x 15 mL |
Pack of 25 Each for $228.53
|
|
|||
Description
Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.
- Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Pouches are 15mL each and are available in 10 packs or 25 packs
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box
Specifications
pH Buffer, individual use pouches | |
pH calibration | |
Liquid | |
Pouch | |
Soluble |
100°C | |
pH Meter and Electrode, single-use | |
Buffer | |
Yes |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.