Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Supplier: Thermo Scientific™ 900023
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
- Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
- Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
Specifications
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution C for General Cleaning, Beaker and Pipet | |
pH Electrode | |
pH Electrode Cleaning Solution | |
30 mL |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.