Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters
Help ensure optimal performance of dissolved oxygen probes with these specialized probe maintenance kits.
Supplier: Thermo Scientific™ 080113
Description
Help ensure optimal performance of Orion polarographic dissolved oxygen probes with Thermo Scientific™ Orion™ Probe Maintenance Kits for Dissolved Oxygen Meters, which include electrolyte solution, polishing disk and two membrane caps.
Warranty: Out of box.Specifications
For Orion 080510MD, 081010MD, 08101 Probes | |
Dissolved Oxygen Probe and Meter | |
Dissolved Oxygen Probe Accessories | |
2 Membrane caps, electrolyte solution, and polishing disk |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.