missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ ROSS™ Sure-Flow™ pH Electrode with Sure-flow junction, BNC connector Edge
Edge

Measure general pH samples as well as viscous or colloidal samples with the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.

Supplier:  Thermo Scientific™ 8172BNWP

Catalog No. 13-642-243


Edge
Explore available promotions

Description

Description

Measure general pH samples as well as viscous or colloidal samples with the free-flowing sleeve junction design of the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.

This general purpose pH electrode offers superior performance for dirty, difficult samples such as soils, sludges, colloids, viscous materials and organics. Junction cleans easily between measurements; pressing down on the cap allows filling solution to flow through the junction carries away any contamination that may interfere with measurement.
  • Material: Glass body
  • pH range: 0 to 14
  • Temperature range: 0° to 100°C
  • Internal reference: ROSS
  • Junction: Sure-Flow
  • Dimensions: 120 × 12mm
  • pH precision: 0.01
  • Fill solution: 3M KCl, Orion Cat. No 810007
  • Cap diameter: 16mm

Includes:

  • Detachable bulb guard, two bottles of Orion ROSS electrode fill solution
Warranty period
12 months from date of purchase
 

Specifications

Specifications

Orion ROSS Sure-Flow pH electrode
Sure-Flow
0 to 14
Combination pH Electrode, glass body, BNC waterproof connector
Biological/Pharmaceutical –TRIS buffer, proteins, enzymes; Emulsions – Foods, cosmetics, oils; Emulsions – Petroleum products, paint; General Purpose – Most sample types; High Ionic Strength – Acids, bases, brines, pH > 12 or pH < 2; Low Ionic Strength –
Detachable bulb guard, 2 x 60mL bottles of ORion ROSS electrode fill solution
0.01
16 mm
WPBNC
Glass Body
0°C to 100°C
3M KCl (810007)
Standard pH Meter, BNC connector
ROSS
32°F to 212°F
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions