Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes for Orion Series Meters
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes.
$865.81 - $1548.62
Specifications
Junction | Glass Fiber |
---|---|
Material | Epoxy Body |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Type | pH/ATC Combination Electrode |
Catalog Number | Mfr. No. | Connector Types | Length (Metric) Cable | For Use With (Equipment) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connector Types | Length (Metric) Cable | For Use With (Equipment) | Price | Quantity | |||
13-643-201
|
Thermo Scientific™
ORI8157BNUMD |
BNC Waterproof, 8-pin MiniDIN | 1 m | Orion Versa Star, Star A and Dual Star Meters |
Each for $1,548.62
|
|
|||
13-642-676
|
Thermo Scientific™
8157BNURCA |
BNC Waterproof, RCA | 1 m | Mettler Toledo Five and Seven series pH meters |
Each for $865.81
|
|
|||
Description
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra pH/ATC electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. They have great stability and drift and come with a two-year replacement warranty.
Measure pH and Temperature with One Electrode
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra electrodes offer the best temperature performance in repeated, varying temperatures of all other electrodes. They have great stability and drift and come with a two-year replacement warranty.
Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.
Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.
Target Markets:
- Pharmaceutical/Biopharmaceutical
- Food Manufacturing
- Academic/Government
- Environmental
Applications:
- Quality Control
- Research
- General Purpose
Specifications
Glass Fiber | |
0 to 14 | |
pH/ATC Combination Electrode | |
ROSS | |
0.03 pH for buffers using automatic temperature compensation | |
2 Years | |
12 mm |
Epoxy Body | |
0°C to 100°C | |
ROSS electrode 3 M KCl fill solution (Cat. No. 810007) | |
7 | |
0.01 | |
120 mm |