Thermo Scientific™ Orion™ Star A Series Portable Meter Protective Armor
Protect your portable meter using Thermo Scientific™ Orion™ Star A Series Portable Meter Protective Armor for use with Thermo Scientific™ Orion™ Star A series portable meters
Supplier:  Thermo Scientific™ STARAAR
Catalog No. 13-645-599
Explore available promotions
Description
Description
Protect your portable meter using Thermo Scientific™ Orion™ Star A Series Portable Meter Protective Armor for use with Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series). Portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electrode holder, conductivity and DO probe holder and pH electrode storage sleeve.
Specifications
Specifications
Star A Protector Armor
Meter Accessories
Orion Star A series portable meter protective armor with hand strap and built-in stand, 810017 pH electrode storage sleeve, STARA-ESPH pH electrode holder, STARA-ESCD conductivity and dissolved oxygen sensor holder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.